siRNA Details
| Gene Symbol: Cgrrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026832.3 | NP_081108.2 | 2 | 246 | ||
| GenBank mRNA | AK004156.1 | - | 2 | 272 | ||
| AK007402.1 | - | 2 | 262 | |||
| AK030403.1 | - | 2 | 247 | |||
| AK030755.1 | - | 2 | 248 | |||
| AK154797.1 | - | 2 | 322 | |||
| BC037677.1 | - | 2 | 265 | 3491666 | ||
| BC114536.1 | - | 1 | 84 | |||

Cgrrf1
Genomic Map