siRNA Details
| Gene Symbol: Ms4a6d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026835.2 | NP_081111.1 | 7 | 817 | ||
| GenBank mRNA | AF237911.1 | - | 7 | 779 | ||
| AK004295.1 | - | 7 | 800 | |||
| AK152859.1 | - | 7 | 798 | |||
| AK156878.1 | - | 7 | 800 | |||
| AY039037.1 | - | 7 | 779 | |||
| BC018331.1 | - | 7 | 754 | 3964037 | ||
| BC029738.1 | - | 7 | 761 | 4016611 | ||

Ms4a6d
Genomic Map