siRNA Details
| Gene Symbol: Pdgfrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026840.3 | NP_081116.3 | 2 | 414 | ||
| GenBank mRNA | AK004179.1 | - | 2 | 306 | ||
| AK081847.1 | - | 2 | 440 | |||
| AK132625.1 | - | 2 | 415 | |||
| BC058275.1 | - | 2 | 406 | 6515674 | ||

Pdgfrl
Genomic Map