siRNA Details
| Gene Symbol: Rgmb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178615.3 | NP_848730.2 | 3 | 1120 | ||
| GenBank mRNA | AJ557514.1 | - | 3 | 1120 | ||
| AK004310.1 | - | 3 | 671 | |||
| AK030386.1 | - | 3 | 1076 | |||
| AK035713.1 | - | 3 | 1067 | |||
| AK047390.1 | - | 3 | 1077 | |||
| AK080819.1 | - | 3 | 1067 | |||
| BC096024.1 | - | 3 | 1067 | 6821755 | ||
| BC138890.1 | - | 3 | 943 | 8861912 | ||

Rgmb
Genomic Map