siRNA Details
| Gene Symbol: Chst8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175140.4 | NP_780349.3 | 2 | 497 | ||
| GenBank mRNA | AB106878.1 | - | 1 | 95 | ||
| AF308145.1 | - | 2 | 315 | |||
| AK005217.1 | - | 2 | 239 | |||
| AK038952.1 | - | 2 | 80 | |||
| AK051290.1 | - | 2 | 79 | |||
| BC051124.1 | - | 2 | 462 | 6528978 | ||
| CF171795.1 | - | 2 | 452 | |||

Chst8
Genomic Map