siRNA Details
| Gene Symbol: Srrm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026886.3 | NP_081162.1 | 1 | 390 | ||
| GenBank mRNA | AK005085.1 | - | 1 | 390 | ||
| AK053836.1 | - | 1 | 122 | |||
| AK165719.1 | - | 1 | 108 | |||
| BC056392.1 | - | 1 | 380 | 6855144 | ||
| JN860221.1 | - | 1 | 37 | |||

Srrm4
Genomic Map