siRNA Details
| Gene Symbol: Phkg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026888.3 | NP_081164.2 | 10 | 1096 | ||
| GenBank mRNA | AK005277.1 | - | 10 | 1041 | ||
| BC138913.1 | - | 10 | 1010 | 8861935 | ||
| BC145734.1 | - | 10 | 1010 | 40131037 | ||
| BQ176174.1 | - | 1 | 545 | |||
| Gene Symbol: Gm166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033040.3 | NP_001028212.1 | 9 | 2138 | ||
| GenBank mRNA | BC066021.1 | - | 9 | 1719 | 5717506 | |

Phkg2
Genomic Map