siRNA Details
| Gene Symbol: Pycr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133705.2 | NP_598466.1 | 7 | 1107 | ||
| GenBank mRNA | AK007535.1 | - | 7 | 1088 | ||
| AK150756.1 | - | 5 | 2227 | |||
| AK152302.1 | - | 5 | 2227 | |||
| AK165618.1 | - | 7 | 1082 | |||
| AK167622.1 | - | 7 | 1063 | |||
| AK179626.1 | - | 2 | 148 | |||
| AK186434.1 | - | 2 | 146 | |||
| BC006882.1 | - | 7 | 1083 | 2647612 | ||

Pycr2
Genomic Map