siRNA Details
| Gene Symbol: Zc3h15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026934.3 | NP_081210.2 | 3 | 325 | ||
| GenBank mRNA | AK005661.1 | - | 3 | 423 | ||
| AK012012.1 | - | 3 | 415 | |||
| AK139511.1 | - | 3 | 420 | |||
| AK152374.1 | - | 3 | 416 | |||
| AK159596.1 | - | 3 | 397 | |||
| AK167696.1 | - | 3 | 418 | |||
| BC031845.1 | - | 3 | 325 | 3983096 | ||
| BC058229.1 | - | 3 | 252 | 6313954 | ||

Zc3h15
Genomic Map