siRNA Details
| Gene Symbol: Zc3h15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026934.3 | NP_081210.2 | 2, 3 | 253 | ||
| GenBank mRNA | AK005661.1 | - | 2, 3 | 351 | ||
| AK012012.1 | - | 2, 3 | 343 | |||
| AK139511.1 | - | 2, 3 | 348 | |||
| AK152374.1 | - | 2, 3 | 344 | |||
| AK159596.1 | - | 2, 3 | 325 | |||
| AK167696.1 | - | 2, 3 | 346 | |||
| BC031845.1 | - | 2, 3 | 253 | 3983096 | ||
| BC058229.1 | - | 2, 3 | 180 | 6313954 | ||

Zc3h15
Genomic Map