siRNA Details
| Gene Symbol: Mcemp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026985.1 | NP_081261.1 | 5, 6 | 495 | ||
| GenBank mRNA | AK007679.1 | - | 5, 6 | 503 | ||
| AK036449.1 | - | 5, 6 | 496 | |||
| AK087121.1 | - | 5, 6 | 501 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mcemp1| Gene Symbol: Mcemp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026985.1 | NP_081261.1 | 5, 6 | 495 | ||
| GenBank mRNA | AK007679.1 | - | 5, 6 | 503 | ||
| AK036449.1 | - | 5, 6 | 496 | |||
| AK087121.1 | - | 5, 6 | 501 | |||