siRNA Details
| Gene Symbol: 3300002I08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027017.1 | NP_081293.4 | 4 | 478 | ||
| GenBank mRNA | AK014369.1 | - | 4 | 478 | ||
| BC132262.1 | - | 4 | 419 | 40130539 | ||
| BC132264.1 | - | 4 | 419 | 40130541 | ||

3300002I08Rik
Genomic Map