siRNA Details
| Gene Symbol: Odf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027019.3 | NP_081295.2 | 6 | 589 | ||
| GenBank mRNA | AB067773.1 | - | 6 | 678 | ||
| AK005883.1 | - | 6 | 700 | |||
| BC104105.1 | - | 6 | 589 | 40043157 | ||
| BC104106.1 | - | 6 | 589 | 40043160 | ||
| BC106806.1 | - | 6 | 589 | 40042632 | ||

Odf3
Genomic Map