siRNA Details
| Gene Symbol: C8g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271777.1 | NP_001258706.1 | 2, 3 | 324 | ||
| NM_027062.2 | NP_081338.1 | 2, 3 | 545 | |||
| GenBank mRNA | AK005962.1 | - | 2, 3 | 328 | ||
| BC019967.1 | - | 2, 3 | 545 | 4023907 | ||
| BY090016.1 | - | 2, 3 | 321 | |||

C8g
Genomic Map