siRNA Details
| Gene Symbol: C8g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271777.1 | NP_001258706.1 | 4 | 439 | ||
| NM_027062.2 | NP_081338.1 | 4 | 660 | |||
| GenBank mRNA | AK005962.1 | - | 4 | 443 | ||
| BC019967.1 | - | 4 | 660 | 4023907 | ||

C8g
Genomic Map