siRNA Details
| Gene Symbol: 1700025G04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197990.3 | NP_932107.1 | 2 | 543 | ||
| GenBank mRNA | AI552192.1 | - | 2 | 192 | ||
| AK006337.1 | - | 2 | 201 | |||
| AK148738.1 | - | 1 | 60 | |||
| AK167392.1 | - | 194 | ||||
| BB840268.1 | - | 2 | 386 | |||
| BC034723.1 | - | 2 | 322 | 3981517 | ||
| BC055912.1 | - | 2 | 358 | 3978655 | ||

1700025G04Rik
Genomic Map