siRNA Details
| Gene Symbol: Klhdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027117.3 | NP_081393.2 | 3, 4 | 656 | ||
| GenBank mRNA | AK009481.1 | - | 3, 4 | 571 | ||
| AK051418.1 | - | 3, 4 | 583 | |||
| AK164223.1 | - | 1, 2 | 32 | |||
| AY513272.1 | - | 3, 4 | 607 | |||
| BC005581.1 | - | 3, 4 | 580 | 3968615 | ||

Klhdc2
Genomic Map