siRNA Details
| Gene Symbol: Cd164l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027152.1 | NP_081428.1 | 2 | 310 | ||
| GenBank mRNA | AK009888.1 | - | 2 | 236 | ||
| AK139809.1 | - | 2 | 311 | |||
| CJ072306.1 | - | 2 | 313 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd164l2| Gene Symbol: Cd164l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027152.1 | NP_081428.1 | 2 | 310 | ||
| GenBank mRNA | AK009888.1 | - | 2 | 236 | ||
| AK139809.1 | - | 2 | 311 | |||
| CJ072306.1 | - | 2 | 313 | |||