siRNA Details
| Gene Symbol: 1810026J23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178619.4 | NP_848734.1 | 2 | 777 | ||
| GenBank mRNA | AK028799.1 | - | 2 | 608 | ||
| AK030602.1 | - | 2 | 630 | |||
| AK034762.1 | - | 2 | 474 | |||
| AK088045.1 | - | 2 | 550 | |||
| AK089121.1 | - | 2 | 536 | |||
| BC025883.1 | - | 2 | 535 | |||
| BC057071.1 | - | 2 | 777 | 5695590 | ||

1810026J23Rik
Genomic Map