siRNA Details
| Gene Symbol: Glod5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027227.2 | NP_081503.1 | 3 | 314 | ||
| GenBank mRNA | AK008008.1 | - | 3 | 317 | ||
| BC061027.1 | - | 3 | 281 | 30312868 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Glod5| Gene Symbol: Glod5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027227.2 | NP_081503.1 | 3 | 314 | ||
| GenBank mRNA | AK008008.1 | - | 3 | 317 | ||
| BC061027.1 | - | 3 | 281 | 30312868 | ||