siRNA Details
| Gene Symbol: Ms4a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023529.2 | NP_076018.2 | 6 | 644 | ||
| GenBank mRNA | AB026046.1 | - | 6 | 653 | ||
| AF237918.1 | - | 6 | 644 | |||
| AK008019.1 | - | 4 | 503 | |||
| BC144935.1 | - | 6 | 641 | 9053483 | ||
| BC150675.1 | - | 6 | 641 | 9087586 | ||

Ms4a10
Genomic Map