siRNA Details
| Gene Symbol: A1cf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081074.1 | NP_001074543.1 | 12 | 1877 | ||
| GenBank mRNA | AY566862.1 | - | 12 | 1874 | ||
| AY566863.1 | - | 12 | 1850 | |||
| BC038942.1 | - | 10 | 1553 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A1cf| Gene Symbol: A1cf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081074.1 | NP_001074543.1 | 12 | 1877 | ||
| GenBank mRNA | AY566862.1 | - | 12 | 1874 | ||
| AY566863.1 | - | 12 | 1850 | |||
| BC038942.1 | - | 10 | 1553 | |||