siRNA Details
| Gene Symbol: Nup43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145706.2 | NP_663752.2 | 8 | 1148 | ||
| GenBank mRNA | AK011422.1 | - | 8 | 1112 | ||
| AK012191.1 | - | 8 | 1142 | |||
| AK090019.1 | - | 8 | 1111 | |||
| AK146262.1 | - | 8 | 1150 | |||
| BC011060.1 | - | 8 | 1136 | 3156436 | ||
| BC052530.1 | - | 8 | 1076 | |||
| DV046453.1 | - | 622 | ||||

Nup43
Genomic Map