siRNA Details
Gene Symbol: Rabep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030566.2 | NP_085043.2 | 9 | 1648 | ||
GenBank mRNA | AK029188.1 | - | 7 | 1006 | ||
AK144836.1 | - | 9 | 1334 | |||
AK157832.1 | - | 9 | 1309 | |||
BC004088.1 | - | 8 | 1394 | 3593722 | ||
BC015287.1 | - | 9 | 1656 | 4240442 |
