siRNA Details
| Gene Symbol: Rabep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030566.2 | NP_085043.2 | 5 | 1119 | ||
| GenBank mRNA | AK029188.1 | - | 3 | 477 | ||
| AK089938.1 | - | 5 | 823 | |||
| AK144836.1 | - | 5 | 805 | |||
| AK157832.1 | - | 5 | 780 | |||
| BC004088.1 | - | 4 | 865 | 3593722 | ||
| BC015287.1 | - | 5 | 1127 | 4240442 | ||

Rabep2
Genomic Map