siRNA Details
Gene Symbol: Dhrs13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183286.2 | NP_899109.2 | 4 | 558 | ||
GenBank mRNA | AK011939.1 | - | 4 | 533 | ||
AK046465.1 | - | 3 | 605 | |||
BB637905.1 | - | 4 | 558 | |||
BC115881.1 | - | 3 | 403 | 40088339 |
