siRNA Details
| Gene Symbol: 5730522E02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080999.1 | - | 3 | 251 | ||
| BY713762.1 | - | 3 | 260 | |||
| BY724063.1 | - | 3 | 253 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5730522E02Rik| Gene Symbol: 5730522E02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080999.1 | - | 3 | 251 | ||
| BY713762.1 | - | 3 | 260 | |||
| BY724063.1 | - | 3 | 253 | |||