siRNA Details
| Gene Symbol: Naa30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081430.1 | NP_001074899.1 | 3 | 1010 | ||
| GenBank mRNA | AK015640.1 | - | 3 | 983 | ||
| AK044235.1 | - | 2 | 630 | |||
| BC054060.1 | - | 1 | 237 | |||
| BC157931.1 | - | 3 | 903 | 9088104 | ||
| BC158065.1 | - | 3 | 903 | 9007364 | ||
| BC172029.1 | - | 3 | 903 | 9054315 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001109099, XM_006251813.

Naa30
Genomic Map