siRNA Details
| Gene Symbol: Ccdc93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025156.2 | NP_001020327.1 | 2 | 205 | ||
| NM_029955.3 | NP_084231.2 | 2 | 205 | |||
| GenBank mRNA | AK014621.1 | - | 2 | 179 | ||
| AK020306.1 | - | 2 | 165 | |||
| AK052300.1 | - | 2 | 170 | |||
| AK082581.1 | - | 2 | 198 | |||
| AK085701.1 | - | 2 | 164 | |||
| AK142302.1 | - | 2 | 223 | |||
| AK159415.1 | - | 2 | 173 | |||
| AK178154.1 | - | 1 | 19 | |||
| BC054083.1 | - | 2 | 108 | 6467830 | ||
| BY136404.1 | - | 2 | 205 | |||
| BY730410.1 | - | 2 | 223 | |||

Ccdc93
Genomic Map