siRNA Details
| Gene Symbol: Cdkl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183294.2 | NP_899117.1 | 9 | 1066 | ||
| GenBank mRNA | AK016781.1 | - | 9 | 1067 | ||
| BC115909.1 | - | 8 | 1014 | 40105122 | ||
| BC118055.1 | - | 8 | 1015 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdkl1| Gene Symbol: Cdkl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183294.2 | NP_899117.1 | 9 | 1066 | ||
| GenBank mRNA | AK016781.1 | - | 9 | 1067 | ||
| BC115909.1 | - | 8 | 1014 | 40105122 | ||
| BC118055.1 | - | 8 | 1015 | |||