siRNA Details
| Gene Symbol: Tmem217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162901.1 | NP_001156373.1 | 3 | 432 | ||
| NM_001162902.1 | NP_001156374.1 | 2 | 340 | |||
| GenBank mRNA | AK016810.1 | - | 3 | 433 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem217| Gene Symbol: Tmem217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162901.1 | NP_001156373.1 | 3 | 432 | ||
| NM_001162902.1 | NP_001156374.1 | 2 | 340 | |||
| GenBank mRNA | AK016810.1 | - | 3 | 433 | ||