siRNA Details
| Gene Symbol: Daw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027725.3 | NP_082001.2 | 7 | 655 | ||
| GenBank mRNA | AK016201.1 | - | 10 | 945 | ||
| AK016977.1 | - | 7 | 656 | |||
| BC049704.1 | - | 7 | 655 | |||
| BC144994.1 | - | 7 | 632 | |||
| BC145995.1 | - | 7 | 655 | 40131298 | ||

Daw1
Genomic Map