siRNA Details
| Gene Symbol: Sdhaf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077713.1 | NP_001071181.1 | 1 | 113 | ||
| GenBank mRNA | AK002229.1 | - | 1 | 95 | ||
| AK004417.1 | - | 1 | 43 | |||
| AK016986.1 | - | 1 | 125 | |||
| AK028031.1 | - | 1 | 84 | |||
| AK046436.1 | - | 128 | ||||
| BC061217.1 | - | 1 | 120 | |||
| BC118927.1 | - | 1 | 113 | 40090186 | ||
| BC118928.1 | - | 1 | 120 | 40090189 | ||

Sdhaf3
Genomic Map