siRNA Details
| Gene Symbol: Tmem80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001141950.1 | NP_001135422.1 | 5 | 418 | ||
| NM_027797.3 | NP_082073.1 | 5 | 487 | |||
| GenBank mRNA | AK017460.1 | - | 5 | 488 | ||
| AK134821.1 | - | 5 | 419 | |||
| AK169083.1 | - | 5 | 377 | |||
| BC022756.1 | - | 5 | 362 | 4922223 | ||

Tmem80
Genomic Map