siRNA Details
| Gene Symbol: Ppp6r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026813.1 | NP_081089.1 | 7, 8 | 1160 | ||
| NM_027805.2 | NP_082081.1 | 7, 8 | 1160 | |||
| GenBank mRNA | AK018403.1 | - | 7, 8 | 1079 | ||
| AK084684.1 | - | 7, 8 | 1161 | |||
| AK173000.1 | - | 1, 2 | 262 | |||
| BC024879.1 | - | 6, 7 | 963 | 4011449 | ||

Ppp6r2
Genomic Map