siRNA Details
| Gene Symbol: Ppp6r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026813.1 | NP_081089.1 | 5 | 692 | ||
| NM_027805.2 | NP_082081.1 | 5 | 692 | |||
| GenBank mRNA | AK018403.1 | - | 5 | 612 | ||
| AK083335.1 | - | 5 | 684 | |||
| AK084684.1 | - | 5 | 693 | |||
| AK209404.1 | - | 2 | 132 | |||
| BC024879.1 | - | 4 | 495 | 4011449 | ||

Ppp6r2
Genomic Map