siRNA Details
| Gene Symbol: Clmp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133733.4 | NP_598494.2 | 4, 5 | 912 | ||
| GenBank mRNA | AB040490.1 | - | 4, 5 | 829 | ||
| AK028983.1 | - | 4, 5 | 882 | |||
| AK033723.1 | - | 4, 5 | 914 | |||
| AK080481.1 | - | 4, 5 | 867 | |||
| AY259213.1 | - | 5, 6 | 541 | |||
| AY326421.1 | - | 4, 5 | 547 | |||
| BC026447.1 | - | 4, 5 | 738 | 4459717 | ||

Clmp
Genomic Map