siRNA Details
| Gene Symbol: Clmp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133733.4 | NP_598494.2 | 6 | 1157 | ||
| GenBank mRNA | AB040490.1 | - | 6 | 1074 | ||
| AK018527.1 | - | 1 | 148 | |||
| AK028983.1 | - | 6 | 1127 | |||
| AK033723.1 | - | 6 | 1159 | |||
| AK080481.1 | - | 6 | 1113 | |||
| AY259213.1 | - | 7 | 786 | |||
| AY326421.1 | - | 6 | 792 | |||
| BC026447.1 | - | 6 | 983 | 4459717 | ||

Clmp
Genomic Map