siRNA Details
| Gene Symbol: Mcm9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027830.2 | NP_082106.2 | 6, 7 | 1374 | ||
| GenBank mRNA | AK018494.1 | - | 5, 6 | 1214 | ||
| AK046636.1 | - | 5, 6 | 1158 | |||
| AK139112.1 | - | 5, 6 | 1221 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mcm9| Gene Symbol: Mcm9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027830.2 | NP_082106.2 | 6, 7 | 1374 | ||
| GenBank mRNA | AK018494.1 | - | 5, 6 | 1214 | ||
| AK046636.1 | - | 5, 6 | 1158 | |||
| AK139112.1 | - | 5, 6 | 1221 | |||