siRNA Details
| Gene Symbol: Mcm9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027830.2 | NP_082106.2 | 3, 4 | 770 | ||
| GenBank mRNA | AK018494.1 | - | 2, 3 | 610 | ||
| AK046636.1 | - | 2, 3 | 554 | |||
| AK139112.1 | - | 2, 3 | 554 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mcm9| Gene Symbol: Mcm9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027830.2 | NP_082106.2 | 3, 4 | 770 | ||
| GenBank mRNA | AK018494.1 | - | 2, 3 | 610 | ||
| AK046636.1 | - | 2, 3 | 554 | |||
| AK139112.1 | - | 2, 3 | 554 | |||