siRNA Details
| Gene Symbol: 9130008F23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027834.3 | NP_082110.1 | 1 | 330 | ||
| GenBank mRNA | AK018601.1 | - | 1 | 331 | ||
| AK033670.1 | - | 1 | 326 | |||
| BC132063.1 | - | 1 | 326 | 40130340 | ||
| BC132065.1 | - | 1 | 326 | 40130342 | ||

9130008F23Rik
Genomic Map