siRNA Details
Gene Symbol: 9130008F23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027834.3 | NP_082110.1 | 1 | 230 | ||
GenBank mRNA | AK018601.1 | - | 1 | 231 | ||
AK033670.1 | - | 1 | 226 | |||
BC132063.1 | - | 1 | 226 | 40130340 | ||
BC132065.1 | - | 1 | 226 | 40130342 |
