siRNA Details
| Gene Symbol: Rnf215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027859.2 | NP_082135.2 | 2 | 438 | ||
| GenBank mRNA | AK002414.1 | - | 2 | 396 | ||
| AK047194.1 | - | 386 | ||||
| AK154140.1 | - | 2 | 421 | |||
| AK183370.1 | - | 1 | 46 | |||
| AK206501.1 | - | 1 | 46 | |||
| BC103627.1 | - | 2 | 362 | 6486672 | ||
| BC115907.1 | - | 2 | 385 | 40105152 | ||
| BC115908.1 | - | 2 | 385 | 40105148 | ||

Rnf215
Genomic Map