siRNA Details
| Gene Symbol: Aox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023617.2 | NP_076106.2 | 23 | 2630 | ||
| GenBank mRNA | AF172276.2 | - | 23 | 2706 | ||
| AK004700.1 | - | 12 | 1426 | |||
| AK149434.1 | - | 23 | 2591 | |||
| AY347569.1 | - | 23 | 2507 | |||
| AY347570.1 | - | 23 | 2507 | |||
| BC049853.1 | - | 23 | 2582 | |||
| BC138876.1 | - | 23 | 2609 | 8861898 | ||

Aox3
Genomic Map