siRNA Details
| Gene Symbol: Dhdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027903.3 | NP_082179.1 | 6 | 851 | ||
| GenBank mRNA | AK005050.2 | - | 6 | 852 | ||
| AK154452.1 | - | 6 | 833 | |||
| BC031710.1 | - | 6 | 829 | |||
| BC116414.2 | - | 6 | 835 | 40103176 | ||

Dhdh
Genomic Map