siRNA Details
| Gene Symbol: Tmem50a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027935.2 | NP_082211.1 | 3, 4 | 237 | ||
| GenBank mRNA | AK014282.1 | - | 3, 4 | 307 | ||
| AK147069.1 | - | 3, 4 | 273 | |||
| AK152630.1 | - | 3, 4 | 321 | |||
| AK152654.1 | - | 3, 4 | 292 | |||
| AK157214.1 | - | 3, 4 | 310 | |||
| AK164989.1 | - | 3, 4 | 303 | |||
| AK165636.1 | - | 3, 4 | 286 | |||
| BC094256.1 | - | 3, 4 | 237 | 6820666 | ||

Tmem50a
Genomic Map