siRNA Details
| Gene Symbol: Syce2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168244.1 | NP_001161716.1 | 3 | 812 | ||
| NM_001168246.1 | NP_001161718.1 | 4 | 511 | |||
| NM_027954.3 | NP_082230.2 | 4 | 493 | |||
| GenBank mRNA | AK005954.1 | - | 4 | 494 | ||
| AK082833.1 | - | 3 | 379 | |||
| AK167332.1 | - | 3 | 811 | |||
| BC094628.1 | - | 4 | 388 | 3374002 | ||
| BC132534.1 | - | 4 | 443 | 40130811 | ||
| BY083329.1 | - | 3 | 225 | |||

Syce2
Genomic Map