siRNA Details
| Gene Symbol: 2310047M10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028005.3 | NP_082281.2 | 1 | 964 | ||
| GenBank mRNA | AK009886.1 | - | 1 | 1007 | ||
| AK151629.1 | - | 1 | 966 | |||
| AK169875.1 | - | 1 | 945 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2310047M10Rik| Gene Symbol: 2310047M10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028005.3 | NP_082281.2 | 1 | 964 | ||
| GenBank mRNA | AK009886.1 | - | 1 | 1007 | ||
| AK151629.1 | - | 1 | 966 | |||
| AK169875.1 | - | 1 | 945 | |||