siRNA Details
| Gene Symbol: Cars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024248.1 | NP_077210.1 | 3 | 368 | ||
| GenBank mRNA | AK009937.1 | - | 3 | 330 | ||
| AK010665.1 | - | 3 | 342 | |||
| AK038968.1 | - | 3 | 381 | |||
| AK041962.1 | - | 3 | 342 | |||
| AK054486.1 | - | 3 | 365 | |||
| CX204497.1 | - | 3 | 369 | |||

Cars2
Genomic Map