siRNA Details
| Gene Symbol: 2410016O06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023633.3 | NP_076122.2 | 1 | 1664 | ||
| GenBank mRNA | AB041553.1 | - | 1 | 1603 | ||
| AK010521.1 | - | 1 | 770 | |||
| AK080870.1 | - | 1 | 1610 | |||
| AK213032.1 | - | 1 | 251 | |||
| BC052152.2 | - | 1 | 1578 | |||
| BC059809.1 | - | 1 | 1615 | |||
| BC138094.1 | - | 1 | 1664 | 8861113 | ||

2410016O06Rik
Genomic Map